missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinic Acetylcholine Receptor beta 4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-60788
This item is not returnable.
View return policy
Description
Nicotinic Acetylcholine Receptor beta 4 Polyclonal specifically detects Nicotinic Acetylcholine Receptor beta 4 in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Nicotinic Acetylcholine Receptor beta 4 | |
| Polyclonal | |
| Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:500 | |
| cholinergic receptor, nicotinic, beta 4, cholinergic receptor, nicotinic, beta polypeptide 4, neuronal acetylcholine receptor subunit beta-4, neuronal nicotinic receptor beta 4 subunit | |
| Rabbit | |
| 52 kDa | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per ml BSA and 50% glycerol with No Preservative | |
| Chrnb4 | |
| Fusion protein from the cytoplasmic loop of the beta 4 subunit of rat nAChR. Accession # Q8R493 | |
| Antigen Affinity-purified | |
| Cellular Markers, Neuronal Cell Markers | |
| 1143 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction