missing translation for 'onlineSavingsMsg'
Learn More

Nicotinic Acetylcholine Receptor beta 4 Antibody (1H11F2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-61743-0.025ml

 View more versions of this product

Product Code. 18651397

  • £173.00 / 0.03mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Nicotinic Acetylcholine Receptor beta 4 Monoclonal specifically detects Nicotinic Acetylcholine Receptor beta 4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nicotinic Acetylcholine Receptor beta 4
Monoclonal
1 mg/mL
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:500
cholinergic receptor, nicotinic, beta 4, cholinergic receptor, nicotinic, beta polypeptide 4, neuronal acetylcholine receptor subunit beta-4, neuronal nicotinic receptor beta 4 subunit
Mouse
56.4 kDa
0.025 mL
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
IgG1
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
1H11F2
Unconjugated
PBS with 0.05% Sodium Azide
CHRNB4
Purified recombinant fragment of human CHRNB4 (AA: extra 22-236) expressed in E. Coli.
Protein G purified
Cellular Markers, Neuronal Cell Markers
1143
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.