missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinic Acetylcholine Receptor beta 2 Antibody (12H2) - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-43573
This item is not returnable.
View return policy
Description
Nicotinic Acetylcholine Receptor beta 2 Monoclonal antibody specifically detects Nicotinic Acetylcholine Receptor beta 2 in Human samples. It is validated for ELISA
Specifications
| Nicotinic Acetylcholine Receptor beta 2 | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA | |
| cholinergic receptor, nicotinic, beta 2 (neuronal), cholinergic receptor, nicotinic, beta polypeptide 2 (neuronal), EFNL3, nAChRB2, neuronal acetylcholine receptor subunit beta-2, neuronal nicotinic acetylcholine receptor beta 2 | |
| The original antibody was generated by immunizing female BALB/c mice with human alpha4beta2 receptor protein purified in n-dodecyl-beta-D-maltopyranoside (DDM). (Uniprot# P17787) | |
| 0.2 mg | |
| Alzheimers Research, Cancer, Endocrinology, Neurodegeneration, Neuroscience | |
| 1141 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 12H2 | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction