missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinic Acetylcholine R alpha 9/CHRNA9 Antibody - BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Nicotinic Acetylcholine R alpha 9/CHRNA9 Polyclonal antibody specifically detects Nicotinic Acetylcholine R alpha 9/CHRNA9 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Nicotinic Acetylcholine R alpha 9/CHRNA9 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:4000 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | cholinergic receptor, nicotinic, alpha 9, cholinergic receptor, nicotinic, alpha polypeptide 9, HSA243342, MGC142109, MGC142135, NACHR alpha 9, NACHR alpha-9, NACHRA9acetylcholine receptor, neuronal nicotinic, alpha-9 subunit, neuronal acetylcholine receptor protein, alpha-9 subunit, neuronal acetylcholine receptor subunit alpha-9, nicotinic acetylcholine receptor subunit alpha 9, Nicotinic acetylcholine receptor subunit alpha-9 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 26-240 of human CHRNA9 (NP_060051.2). ADGKYAQKLFNDLFEDYSNALRPVEDTDKVLNVTLQITLSQIKDMDERNQILTAYLWIRQIWHDAYLTWDRDQYDGLDSIRIPSDLVWRPDIVLYNKADDESSEPVNTNVVLRYDGLITWDAPAITKSSCVVDVTYFPFDNQQCNLTFGSWTYNGNQVDIFNALDSGDLSDFIEDVEWEVHGMPAVKNVISYGCCSEPYPDVTFTLLLKRRSSFY |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?