missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinic Acetylcholine R alpha 5/CHRNA5 Antibody (8F11G8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61678
This item is not returnable.
View return policy
Description
Nicotinic Acetylcholine R alpha 5/CHRNA5 Monoclonal specifically detects Nicotinic Acetylcholine R alpha 5/CHRNA5 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| Nicotinic Acetylcholine R alpha 5/CHRNA5 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
| Cholinergic receptor, neuronal nicotinic, alpha polypeptide-5, cholinergic receptor, nicotinic, alpha 5, cholinergic receptor, nicotinic, alpha polypeptide 5, LNCR2, NACHRA5, neuronal acetylcholine receptor subunit alpha-5, neuronal nicotinic acetylcholine receptor, alpha5 subunit | |
| Mouse | |
| 53 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| 8F11G8 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| CHRNA5 | |
| Purified recombinant fragment of human CHRNA5 (AA: extra 23-254) expressed in E. Coli. | |
| Protein G purified | |
| Signal Transduction | |
| 1138 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction