missing translation for 'onlineSavingsMsg'
Learn More

Nicotinic Acetylcholine R alpha 5/CHRNA5 Antibody (7D3), Novus Biologicals™

Product Code. 18395017 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18395017 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18395017 Supplier Novus Biologicals Supplier No. H00001138M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Nicotinic Acetylcholine R alpha 5/CHRNA5 Monoclonal antibody specifically detects Nicotinic Acetylcholine R alpha 5/CHRNA5 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nicotinic Acetylcholine R alpha 5/CHRNA5
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 7D3
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_000736
Gene Alias Cholinergic receptor, neuronal nicotinic, alpha polypeptide-5, cholinergic receptor, nicotinic, alpha 5, cholinergic receptor, nicotinic, alpha polypeptide 5, LNCR2, NACHRA5, neuronal acetylcholine receptor subunit alpha-5, neuronal nicotinic acetylcholine receptor, alpha5 subunit
Host Species Mouse
Immunogen CHRNA5 (NP_000736, 38 a.a. ~ 131 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SEPSSIAKHEDSLLKDLFQDYERWVRPVEHLNDKIKIKFGLAISQLVDVDEKNQLMTTNVWLKQEWIDVKLRWNPDDYGGIKVIRVPSDSVWTP
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1138
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.