missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinic Acetylcholine R alpha 5/CHRNA5 Antibody (2D2D10) - BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Nicotinic Acetylcholine R alpha 5/CHRNA5 Monoclonal specifically detects Nicotinic Acetylcholine R alpha 5/CHRNA5 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
Specifications
| Antigen | Nicotinic Acetylcholine R alpha 5/CHRNA5 |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Clone | 2D2D10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | Cholinergic receptor, neuronal nicotinic, alpha polypeptide-5, cholinergic receptor, nicotinic, alpha 5, cholinergic receptor, nicotinic, alpha polypeptide 5, LNCR2, NACHRA5, neuronal acetylcholine receptor subunit alpha-5, neuronal nicotinic acetylcholine receptor, alpha5 subunit |
| Gene Symbols | CHRNA5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?