missing translation for 'onlineSavingsMsg'
Learn More

Nicotinic Acetylcholine R alpha 4/CHRNA4 Antibody (4E3), Novus Biologicals™

Product Code. 18332448 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18332448 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18332448 Supplier Novus Biologicals Supplier No. H00001137M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Nicotinic Acetylcholine R alpha 4/CHRNA4 Monoclonal antibody specifically detects Nicotinic Acetylcholine R alpha 4/CHRNA4 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nicotinic Acetylcholine R alpha 4/CHRNA4
Applications Western Blot, ELISA
Classification Monoclonal
Clone 4E3
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_000735
Gene Alias BFNC, cholinergic receptor, nicotinic, alpha 4, cholinergic receptor, nicotinic, alpha polypeptide 4, EBN, EBN1, ENFL1, FLJ95812, NACHR, NACHRA4, NACRA4, neuronal acetylcholine receptor subunit alpha-4, neuronal nicotinic acetylcholine receptor alpha-4 subunit
Host Species Mouse
Immunogen CHRNA4 (NP_000735, 29 a.a. ~ 128 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. HVETRAHAEERLLKKLFSGYNKWSRPVANISDVVLVRFGLSIAQLIDVDEKNQMMTTNVWVKQEWHDYKLRWDPADYENVTSIRIPSELIWRPDIVLYNN
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1137
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.