missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinic Acetylcholine R alpha 10/CHRNA10 Antibody (4E2D1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61666-0.025ml
This item is not returnable.
View return policy
Description
Nicotinic Acetylcholine R alpha 10/CHRNA10 Monoclonal specifically detects Nicotinic Acetylcholine R alpha 10/CHRNA10 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| Nicotinic Acetylcholine R alpha 10/CHRNA10 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
| cholinergic receptor, nicotinic, alpha 10, cholinergic receptor, nicotinic, alpha polypeptide 10, NACHR alpha-10, NACHRA10, neuronal acetylcholine receptor subunit alpha-10, Nicotinic acetylcholine receptor subunit alpha-10 | |
| Mouse | |
| 49.7 kDa | |
| 0.025 mL | |
| 57053 | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| 4E2D1 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| CHRNA10 | |
| Purified recombinant fragment of human CHRNA10 (AA: extra 25-237) expressed in E. Coli. | |
| Protein G purified | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction