missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinic Acetylcholine R alpha 1/CHRNA1 Antibody (mAb 192), Alexa Fluor™ 647, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Nicotinic Acetylcholine R alpha 1/CHRNA1 Monoclonal specifically detects Nicotinic Acetylcholine R alpha 1/CHRNA1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | Nicotinic Acetylcholine R alpha 1/CHRNA1 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | mAb 192 |
| Conjugate | Alexa Fluor 647 |
| Dilution | Western Blot, ELISA, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | acetylcholine receptor subunit alpha, ACHRA, ACHRD, CHNRA, cholinergic receptor, nicotinic, alpha 1 (muscle), CHRNA, CMS2A, FCCMS, nicotinic acetylcholine receptor alpha subunit, nicotinic cholinergic receptor alpha 1, nicotinic, alpha polypeptide 1 (muscle), SCCMS |
| Gene Symbols | CHRNA1 |
| Host Species | Rat |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?