missing translation for 'onlineSavingsMsg'
Learn More

Nicotinic Acetylcholine R alpha 1/CHRNA1 Antibody (mAb 192), Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18355201 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18355201 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18355201 Supplier Novus Biologicals Supplier No. NBP281070AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Nicotinic Acetylcholine R alpha 1/CHRNA1 Monoclonal specifically detects Nicotinic Acetylcholine R alpha 1/CHRNA1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nicotinic Acetylcholine R alpha 1/CHRNA1
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone mAb 192
Conjugate Alexa Fluor 488
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias acetylcholine receptor subunit alpha, ACHRA, ACHRD, CHNRA, cholinergic receptor, nicotinic, alpha 1 (muscle), CHRNA, CMS2A, FCCMS, nicotinic acetylcholine receptor alpha subunit, nicotinic cholinergic receptor alpha 1, nicotinic, alpha polypeptide 1 (muscle), SCCMS
Gene Symbols CHRNA1
Host Species Rat
Immunogen Human muscle AChR.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1134
Test Specificity The antibody binds to human muscle AChR with a Kd of 10 pM, to mouse muscle AChR with a Kd of 50 pM, and to rat muscle AChR with a Kd of 646 nM.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.