missing translation for 'onlineSavingsMsg'
Learn More

Nicotine Bitartrate Dihydrate, MP Biomedicals™

Product Code. 11482902 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11482902 250 mg 250mg
11492902 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11482902 Supplier MP Biomedicals Supplier No. 0215355483
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6-3-6019 | C18H30N2O14 | 498.44 g/mol

Chemical Identifiers

Molecular Formula C18H30N2O14
Molecular Weight (g/mol) 498.44
MDL Number MFCD00150899
InChI Key LDMPZNTVIGIREC-HYRPJEHINA-N
SMILES O.O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.CN1CCC[C@H]1C1=CN=CC=C1

Specifications

CAS 6-3-6019
Melting Point 93°C to 95°C (literature)
Color White
pH 3.18 (10% aq. soln.) (literature)
Quantity 250 mg
Synonym nicotine ditartrate dihydrate, unii-r7m676m8yv, nicotine bitartrate usan, 2r,3r-2,3-dihydroxybutanedioic acid;3-2s-1-methylpyrrolidin-2-yl pyridine;dihydrate, nicotine bitartrate who-dd, nicotine acid tartrate dihydrate, s-3-1-methylpyrrolidin-2-yl pyridine bis 2r,3r-2,3-dihydroxysuccinate dihydrate, 2r,3r-2,3-dihydroxybutanedioic acid; 3-2s-1-methylpyrrolidin-2-yl pyridine; dihydrate, 3-2s-1-methyl-2-pyrrolidinyl pyridine, 2r,3r-2,3-dihydroxybutanedioate 1:2 , dihydrate
Solubility Information Soluble in water (50mg/mL - clear to slightly hazy, colorless to faint yellow solution) or alcohol.
IUPAC Name bis((2R,3R)-2,3-dihydroxybutanedioic acid) 3-[(2S)-1-methylpyrrolidin-2-yl]pyridine dihydrate
Formula Weight 498.44
Physical Form Powder
Chemical Name or Material Nicotine Bitartrate Dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.