missing translation for 'onlineSavingsMsg'
Learn More

(-)-Nicotine Ditartrate, TRC

Product Code. 30040446
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30040446 100 mg 100mg
30041022 1 g 1g
30007704 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30040446 Supplier Toronto Research Chemicals Supplier No. TRCN424598100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-31-6
Molecular Formula C10 H14 N2 . 2 C4 H6 O6
Molecular Weight (g/mol) 462.41
Synonym Nicotine ditartrate, 3-[(2S)-1-Methyl-2-pyrrolidinyl]pyridine (2R,3R)-2,3-dihydroxybutanedioate (1:2), Nicotine tartrate (1:2), (S)-3-(1-Methyl-2-pyrrolidinyl)pyridine [R-(R*,R*)]-2,3-dihydroxybutanedioate (1:2), (-)-Nicotine ditartrate, (-)-Nicotine hydrogen (+)-tartrate, NSC 97238, Nicotine acid tartrate, Nicotine bitartrate, Nicotine hydrogen (+)-tartrate, Nicotine hydrogen tartrate, Nicotine tartrate, S-(-)-Nicotine hydrogen-(+)-tartrate
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid;3-[(2S)-1-methylpyrrolidin-2-yl]pyridine
SMILES CN1CCC[C@H]1c2cccnc2.O[C@H]([C@@H](O)C(=O)O)C(=O)O.O[C@H]([C@@H](O)C(=O)O)C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H14N2.2C4H6O6/c1-12-7-3-5-10(12)9-4-2-6-11-8-9;2*5-1(3(7)8)2(6)4(9)10/h2,4,6,8,10H,3,5,7H2,1H3;2*1-2,5-6H,(H,7,8)(H,9,10)/t10-;2*1-,2-/m011/s1
Formula Weight 462.1486
Percent Purity >95%
Chemical Name or Material Nicotine ditartrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.