missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ (-)-Nicotine ditartrate
Description
Nicotinic acetylcholine receptor (nAChR) agonist (Ki values are 1, > 1000, 4000 and 7130 nM at α4β2, α1β1δγ, rat α7 and human α7 respectively). Exhibits vasoconstrictive, hypertensive and prothrombotic activity in vivo.
Specifications
Specifications
| CAS | 65-31-6 |
| Quantity | 50 mg |
| Molecular Formula | C18H26N2O12 |
| Formula Weight | 462.41 |
| Purity | >99% |
| Molecular Weight (g/mol) | 462.408 |
| Chemical Name or Material | (-)-Nicotine ditartrate |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?