missing translation for 'onlineSavingsMsg'
Learn More

Nicotinamide, TRC

Product Code. 30133058
Change view
Click to view available options
Quantity:
50 mg
1 g
100 g
Unit Size:
100g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133058 100 g 100g
17972125 1 g 1g
17950885 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133058 Supplier Toronto Research Chemicals Supplier No. TRCN407750100G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-92-0
Molecular Formula C6 H6 N2 O
Molecular Weight (g/mol) 122.12
Synonym Nicotinamide, 3-Pyridinecarboxamide, 3-(Aminocarbonyl)pyridine, 3-Amidopyridine, 3-Carbamoylpyridine, 3-Pyridinecarboxylic acid amide, Aminicotin, Benicot, Delonin Amide, Dipegyl, NAM, NSC 13128, NSC 27452, Niacinamide, Niavit PP, Nicamina, Nicamindon, Nicasir, Nicobion, Nicofort, Nicosan 2, Nicosylamide, Nicotilamide, Nicotine acid amide, Nicotinic acid amide, Nicotinic amide, Nicotylamide, Nicovit, Nicovitina, Nictoamide, Niocinamide, Niozymin, Papulex, Pelmin, Pelmine, Pelonin amide, Vi-Nicotyl, Vitamin B, Vitamin B3, m-(Aminocarbonyl)pyridine, β-Pyridinecarboxamide
IUPAC Name pyridine-3-carboxamide
SMILES NC(=O)c1cccnc1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C6H6N2O/c7-6(9)5-2-1-3-8-4-5/h1-4H,(H2,7,9)
Formula Weight 122.048
Percent Purity >95%
Chemical Name or Material Nicotinamide (Vitamin B3)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.