missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Nicotinamide Riboside
GREENER_CHOICE

Product Code. 18795636
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. Quantity unitSize
18795636 50 mg 50mg
1 options
This item is not returnable. View return policy

Product Code. 18795636

Brand: Tocris Bioscience™ 6018/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

NAD+ precursor; substrate for nicotinamide riboside kinases

Nicotinamide Riboside is an NAD+ precursor. It is a substrate for nicotinamide riboside kinases (NRK1/2). Nicotinamide Riboside shows neuroprotective effects in a mouse model of type 2 diabetes and improves mitochondrial function in muscle stem cells in aged mice. The compound also corrects non-alchoholic fatty liver disease phenotype induced by NAD+ deficiency or high-fat diet in mice. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(Aminocarbonyl)-1-β-D-ribofuranosylpyridinium chloride
CAS 23111-00-4
Quantity 50 mg
Target Enzyme Substrates
Molecular Formula C11H15ClN2O5
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.