missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinamide N-Methyltransferase/NNMT Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-03406-100UL
This item is not returnable.
View return policy
Description
Nicotinamide N-Methyltransferase/NNMT Polyclonal specifically detects Nicotinamide N-Methyltransferase/NNMT in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated.
Specifications
| Nicotinamide N-Methyltransferase/NNMT | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 4837 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence, KnockDown | |
| Unconjugated | |
| EC 2.1.1.1, nicotinamide N-methyltransferase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-264 of human Nicotinamide N-Methyltransferase/NNMT (NP_006160.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction