missing translation for 'onlineSavingsMsg'
Learn More

Nicotinamide N-Methyltransferase/NNMT Antibody (954208) [mFluor Violet 500 SE], Novus Biologicals™

Artikelnummer. 30735839 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Quantity:
0.1 mL
Packungsgröße:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
30735839 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30735839 Lieferant Novus Biologicals Lieferanten-Nr. FAB7736MFV500

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Mouse Monoclonal Antibody

Nicotinamide N-Methyltransferase/NNMT Monoclonal antibody specifically detects Nicotinamide N-Methyltransferase/NNMT in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen Nicotinamide N-Methyltransferase/NNMT
Applications Western Blot
Classification Monoclonal
Clone 954208
Conjugate mFluor Violet 500 SE
Formulation 50mM Sodium Borate
Gene Alias EC 2.1.1.1, nicotinamide N-methyltransferase
Host Species Mouse
Immunogen E.coli-derived recombinant human Nicotinamide N-Methyltransferase/NNMT, Met1-Leu264, Accession # P40261
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4837
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Mehr anzeigen Weniger anzeigen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.