Learn More
β-Nicotinamide Mononucleotide Mouse, anti-Human, Clone: 3A1F11, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | β-Nicotinamide Mononucleotide |
|---|---|
| Clone | 3A1F11 |
| Concentration | 1000 μg/mL |
| Applications | ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18068955
|
Proteintech
68638-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18078955
|
Proteintech
68638-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The β-Nicotinamide Mononucleotide antibody from Proteintech is a mouse monoclonal antibody generated with ptg of na β-Nicotinamide Mononucleotide. This antibody recognizes human antigen. The β-Nicotinamide Mononucleotide antibody has been validated for the following applications: ELISA analysis.
Specifications
| β-Nicotinamide Mononucleotide | |
| 1000 μg/mL | |
| Liquid | |
| Human | |
| IgG1 | |
| Store at -20°C. Stable for one year after shipment. |
| 3A1F11 | |
| ELISA | |
| Mouse | |
| NMN, β NMN, β-Nicotinamide Mononucleotide, β-NMN | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.