missing translation for 'onlineSavingsMsg'
Learn More

β-Nicotinamide mononucleotide, MedChemExpress

Product Code. 16478778
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478778 10 mg 10mg
16474697 50 mg 50mg
16468926 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16478778 Supplier Medchem Express Supplier No. HYF0004
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress β-nicotinamide mononucleotide (β-NM) is a product of the nicotinamide phosphoribosyltransferase (NAMPT) reaction and a key NAD+ intermediate. The pharmacological activities of β-nicotinamide mononucleotide include its role in cellular biochemical functions, cardioprotection, diabetes, Alzheimer's disease, and complications associated with obesity.

β-nicotinamide mononucleotide (β-NM) is a product of the nicotinamide phosphoribosyltransferase (NAMPT) reaction and a key NAD+ intermediate. The pharmacological activities of β-nicotinamide mononucleotide include its role in cellular biochemical functions, cardioprotection, diabetes, Alzheimer's disease, and complications associated with obesity.

Specifications

Chemical Name or Material β-Nicotinamide mononucleotide
CAS 1094-61-7
Color White
Purity Grade Notes Research
Molecular Formula C11H15N2O8P
Quantity 10 mg
Synonym β-NM NMN
Solubility Information H2O : 83.33 mg/mL (249.33 mM; Need ultrasonic)
SMILES NC(C1=C[N+]([C@@H]2O[C@H](COP(O)([O-])=O)[C@@H](O)[C@H]2O)=CC=C1)=O
Molecular Weight (g/mol) 334.22
Formula Weight 334.22
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.