missing translation for 'onlineSavingsMsg'
Learn More

Nicotinamide, MedChemExpress

Product Code. 16492059
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492059 1 g 1g
16480788 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16492059 Supplier Medchem Express Supplier No. HYB0150
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nicotinamide is a form of vitamin B3 or niacin that inhibits sirtuin 2 (SIRT2) activity in vitro, with an EC50 of 2 μM. Nicotinamide inhibits up to 90% melanoma cell number and increases cellular NAD+, ATP, ROS levels. Nicotinamide inhibts tumor growth in vivo and improves survival of melanoma-bearing mice, which can be used for the research of skin cancers such as melanoma.

Nicotinamide is a form of vitamin B3 or niacin that inhibits sirtuin 2 (SIRT2) activity in vitro, with an EC50 of 2 μM. Nicotinamide inhibits up to 90% melanoma cell number and increases cellular NAD+, ATP, ROS levels. Nicotinamide inhibts tumor growth in vivo and improves survival of melanoma-bearing mice, which can be used for the research of skin cancers such as melanoma.

Specifications

Chemical Name or Material Nicotinamide
CAS 98-92-0
Color White
Purity Grade Notes Research
Molecular Formula C6H6N2O
Quantity 1 g
Synonym Niacinamide Nicotinic acid amide
Solubility Information DMSO : 50 mg/mL (409.43 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (409.43 mM)
SMILES O=C(C1=CC=CN=C1)N
Molecular Weight (g/mol) 122.12
Formula Weight 122.12
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.