missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicorandil-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Nicorandil D4 (pyridyl-D4) |
| CAS | 1132681-23-2 |
| Molecular Formula | C8 2H4 H5 N3 O4 |
| Quantity | 10 mg |
| Synonym | 3-Pyridine-2,4,5,6-d4-carboxamide','N-[2-(nitrooxy)ethyl]-, Nicorandil-d4, Nicorandil D4 (pyridyl-D4) |
| InChI Formula | InChI=1S/C8H9N3O4/c12-8(7-2-1-3-9-6-7)10-4-5-15-11(13)14/h1-3,6H,4-5H2,(H,10,12)/i1D,2D,3D,6D |
| SMILES | [2H]c1nc([2H])c(C(=O)NCCO[N+](=O)[O-])c([2H])c1[2H] |
| IUPAC Name | 2-[(2,4,5,6-tetradeuteriopyridine-3-carbonyl)amino]ethyl nitrate |
| Molecular Weight (g/mol) | 215.2 |
| Formula Weight | 215.0844 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?