missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NICN1 Polyclonal Antibody

Product Code. 15978844
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15978844 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15978844 Supplier Invitrogen™ Supplier No. PA566124

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PSVTFPKWLSH PVPCEQPALL REGLPDPSRV SSEVQQMWAL TEMIRASHTS ARIGRFDVDG CYDLNLLSYT Highest antigen sequence identity to the following orthologs - mouse 89%, rat 87%.

This protein encoded by this gene localizes to the nucleus and is expressed in numerous tissues including brain, testis, liver, and kidney. Computer predictions indicate that this region of the 3' UTR contains hairpin-forming self-complementary sequence which is possibly excised after transcription. This gene has a pseudogene on chromosome X.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NICN1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NICN1
Gene Accession No. Q9BSH3
Gene Alias NICN1; nicolin 1; nicolin-1; NPCEDRG; PGs5; Tubulin polyglutamylase complex subunit 5
Gene Symbols NICN1
Host Species Rabbit
Immunogen Recombinant Human NICN1. Recombinant protein control fragment (Product #RP-105090).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84276
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.