missing translation for 'onlineSavingsMsg'
Learn More

Niclosamide, TRC

Product Code. 30475590
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30475590 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30475590 Supplier Toronto Research Chemicals Supplier No. TRCN395500500MG
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Niclosamide
CAS 50-65-7
Purity Grade Notes HPLC
Molecular Formula C13 H8 Cl2 N2 O4
Quantity 500 mg
Synonym Niclosamide anhydrous, 5-Chloro-N-(2-chloro-4-nitrophenyl)-2-hydroxybenzamide, Benzamide','5-chloro-N-(2-chloro-4-nitrophenyl)-2-hydroxy-Salicylanilide','2',5-dichloro-4'-nitro- (8CI), 2-Chloro-4-nitrophenylamide-6-chlorosalicylic acid, 2-Hydroxy-5-chloro-N-(2-chloro-4-nitrophenyl)benzamide, 2',5-Dichloro-4'-nitrosalicylanilide, 5-Chloro-2'-chloro-4'-nitrosalicylanilide, 5-Chloro-N-(2-chloro-4-nitrophenyl)-2-hydroxybenzamide, 5-Chloro-N-(2'-chloro-4'-nitrophenyl)salicylamide, BAY 2353, Bayer 2353, Bayluscid, Cestocid, Cestocide, Devermin, Devermine, Fedal-Telmin, Fenasal, HL 2447, Helmiantin, Iomesan, Lintex, Mansonil, Mato, N-(2-Chloro-4-nitrophenyl)-5-chlorosalicylamide, N-(2'-Chloro-4'-nitrophenyl)-5-chlorosalicylamide, NSC 178296, Nasemo, Niclocide, Niclosamide, Phenasal, Radeverm, Ruby, Sagimid, Sulqui, Tredemine, Utosamide, Vermitid, Vermitin, WR 46234, Yomesan, Zestocarp
InChI Formula InChI=1S/C13H8Cl2N2O4/c14-7-1-4-12(18)9(5-7)13(19)16-11-3-2-8(17(20)21)6-10(11)15/h1-6,18H,(H,16,19)
SMILES Oc1ccc(Cl)cc1C(=O)Nc2ccc(cc2Cl)[N+](=O)[O-]
IUPAC Name 5-chloro-N-(2-chloro-4-nitrophenyl)-2-hydroxybenzamide
Molecular Weight (g/mol) 327.12
Formula Weight 325.9861
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.