missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Niclosamide-olamine 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Niclosamide-olamine |
| CAS | 1420-04-8 |
| Analyte or Component Names | Niclosamide-olamine |
| Shipping Condition | +20°C |
| Molecular Formula | C13 H8 Cl2 N2 O4 . C2 H7 N O |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Salicylanilide, 2',5-dichloro-4'-nitro-, compd. with 2-aminoethanol (1:1) (8CI), Ethanol, 2-amino-, compd. with 2',5-dichloro-4'-nitrosalicylanilide (1:1) (8CI), Ethanol, 2-amino-, compd. with 5-chloro-N-(2-chloro-4-nitrophenyl)-2-hydroxybenzamide (1:1) (9CI), 2',5-Dichloro-4'-nitrosalicylanilide, 2-aminoethanol salt, BAY 6076, BAY 73, Bayer 6076, Bayer 73, Baylucit, Bayluscide, Clonitralid, Clonitralide, Clonitrilide, HL 2448, M 73, Mollutox, Niclosamide ethanolamine salt, Niclosamide-olamine, Phenasal ethanolamine salt, SR 73, Uccmaluscide |
| InChI Formula | InChI=1S/C13H8Cl2N2O4.C2H7NO/c14-7-1-4-12(18)9(5-7)13(19)16-11-3-2-8(17(20)21)6-10(11)15;3-1-2-4/h1-6,18H,(H,16,19);4H,1-3H2 |
| SMILES | NCCO.Oc1ccc(Cl)cc1C(=O)Nc2ccc(cc2Cl)[N+](=O)[O-] |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?