missing translation for 'onlineSavingsMsg'
Learn More

Niclosamide, MedChemExpress

Product Code. 16440958
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16440958 5 g 5g
16418508 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16440958 Supplier Medchem Express Supplier No. HYB0497
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Niclosamide (BAY2353) is an orally active antihelminthic agent used in parasitic infection research. Niclosamide is a STAT3 inhibitor with an IC50 of 0.25 μM in HeLa cells. Niclosamide has biological activities against cancer, inhibits DNA replication in Vero E6 cells.

Niclosamide (BAY2353) is an orally active antihelminthic agent used in parasitic infection research. Niclosamide is a STAT3 inhibitor with an IC50 of 0.25 μM in HeLa cells. Niclosamide has biological activities against cancer, inhibits DNA replication in Vero E6 cells.

Specifications

Chemical Name or Material Niclosamide
Physical Form Powder
CAS 50-65-7
Color Off-White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C13H8Cl2N2O4
For Use With (Application) COVID-19-immunoregulation
Synonym BAY2353
Solubility Information DMF : 5 mg/mL (15.28 mM; Need ultrasonic) ∣DMSO : 4.55 mg/mL (13.91 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(NC1=CC=C([N+]([O-])=O)C=C1Cl)C2=CC(Cl)=CC=C2O
Molecular Weight (g/mol) 327.12
Formula Weight 327.12
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.