missing translation for 'onlineSavingsMsg'
Learn More

Niclosamide Ethanolamine Salt, TRC

Product Code. 17953935
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17953935 50 mg 50mg
17923485 1 g 1g
17902025 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17953935 Supplier Toronto Research Chemicals Supplier No. TRCN39550550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1420-04-8
Molecular Formula C13 H8 Cl2 N2 O4 . C2 H7 N O
Molecular Weight (g/mol) 388.2
Synonym Salicylanilide, 2',5-dichloro-4'-nitro-, compd. with 2-aminoethanol (1:1) (8CI), Ethanol, 2-amino-, compd. with 2',5-dichloro-4'-nitrosalicylanilide (1:1) (8CI), Ethanol, 2-amino-, compd. with 5-chloro-N-(2-chloro-4-nitrophenyl)-2-hydroxybenzamide (1:1) (9CI), 2',5-Dichloro-4'-nitrosalicylanilide, 2-aminoethanol salt, BAY 6076, BAY 73, Bayer 6076, Bayer 73, Baylucit, Bayluscide, Clonitralid, Clonitralide, Clonitrilide, HL 2448, M 73, Mollutox, Niclosamide ethanolamine salt, Niclosamide-olamine, Phenasal ethanolamine salt, SR 73, Uccmaluscide
IUPAC Name 2-aminoethanol;5-chloro-N-(2-chloro-4-nitrophenyl)-2-hydroxybenzamide
SMILES NCCO.Oc1ccc(Cl)cc1C(=O)Nc2ccc(cc2Cl)[N+](=O)[O-]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H8Cl2N2O4.C2H7NO/c14-7-1-4-12(18)9(5-7)13(19)16-11-3-2-8(17(20)21)6-10(11)15;3-1-2-4/h1-6,18H,(H,16,19);4H,1-3H2
Formula Weight 387.0389
Chemical Name or Material Niclosamide-olamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.