missing translation for 'onlineSavingsMsg'
Learn More

Niclosamide, 97+%, Thermo Scientific Chemicals

Product Code. 15712987 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15712987 100 g 100g
15702987 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15712987 Supplier Thermo Scientific Acros Supplier No. 461841000
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-65-7
Molecular Formula C13H8Cl2N2O4
Molecular Weight (g/mol) 327.12
MDL Number MFCD00057597
InChI Key RJMUSRYZPJIFPJ-UHFFFAOYSA-N
Synonym niclosamide, 5-chloro-n-2-chloro-4-nitrophenyl-2-hydroxybenzamide, niclocide, bayluscid, phenasal, tredemine, fenasal, yomesan, dichlosale, helmiantin
PubChem CID 4477
IUPAC Name 5-chloro-N-(2-chloro-4-nitrophenyl)-2-hydroxybenzamide
SMILES OC1=CC=C(Cl)C=C1C(=O)NC1=CC=C(C=C1Cl)[N+]([O-])=O

Specifications

Melting Point 225.0°C to 230.0°C
Assay Percent Range 97% min. (HPLC)
Quantity 100 g
Solubility Information Solubility in water: practically insoluble. Other solubilities: sparingly soluble in ethanol, chloroform, ether
Formula Weight 327.12
Percent Purity 97+%
Chemical Name or Material Niclosamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.