missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel, Weld Filler, Alloy 244H, N02216, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Nickel, Weld Filler, Alloy 244H, N02216, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Antimony/Tin/Zirconium/Arsenic/Oxygen/Magnesium/Boron/Molybdenum/Phosphorus/Vanadium/Cobalt/Nitrogen/Copper/Carbon/Silicon/Titanium/Aluminium/Niobium/Chromium/Manganese/Iron/Nickel/Barium/Beryllium/Bromine/Cerium/Cesium/Chlorine/Dysprosium/Erbium/Europium/Fluorine/Gadolinium/Gold/Holmium/Indium/Iodine/Iridium/Lithium/Lutetium/Mercury/Neodymium/Osmium/Platinum/Potassium/Praseodymium/Rhodium/Rubidium/Ruthenium/Scandium/Sodium/Hydrogen/Zinc/Lanthanum/Silver/Gallium/Palladium/Samarium/Selenium/Tellurium/Terbium/Thallium/Thorium/Thulium/Uranium/Ytterbium/Yttrium/Strontium/Bismuth/Cadmium/Hafnium/Rhenium/Lead/Calcium/Germanium/Tantalum/Tungsten |
| CAS | 7440-02-0 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Antimony 0.00012 %/Tin 0.0003 %/Zirconium 0.0008 %/Arsenic 0.0009 %/Oxygen 0.0009 %/Magnesium 0.0014 %/Boron 0.002 %/Molybdenum 0.003 %/Phosphorus 0.003 %/Vanadium 0.004 %/Cobalt 0.005 %/Nitrogen 0.0087 %/Copper 0.013 %/Carbon 0.052 %/Silicon 0.16 %/Titanium 0.2 %/Aluminium 0.23 %/Niobium 0.7 %/Chromium 6.9 %/Manganese 10.9 %/Iron 39.9 %/Nickel 41.1 %/Barium (<0.0001) %/Beryllium (<0.0001) %/Bromine (<0.0001) %/Cerium (<0.0001) %/Cesium (<0.0001) %/Chlorine (<0.0001) %/Dysprosium (<0.0001) %/Erbium (<0.0001) %/Europium (<0.0001) %/Fluorine (<0.0001) %/Gadolinium (<0.0001) %/Gold (<0.0001) %/Holmium (<0.0001) %/Indium (<0.0001) %/Iodine (<0.0001) %/Iridium (<0.0001) %/Lithium (<0.0001) %/Lutetium (<0.0001) %/Mercury (<0.0001) %/Neodymium (<0.0001) %/Osmium (<0.0001) %/Platinum (<0.0001) %/Potassium (<0.0001) %/Praseodymium (<0.0001) %/Rhodium (<0.0001) %/Rubidium (<0.0001) %/Ruthenium (<0.0001) %/Scandium (<0.0001) %/Sodium (<0.0001) %/Hydrogen (<0.0003) %/Zinc (<0.0003) %/Lanthanum (<0.0005) %/Silver (<0.0005) %/Gallium (<0.0010) %/Palladium (<0.0010) %/Samarium (<0.0010) %/Selenium (<0.0010) %/Tellurium (<0.0010) %/Terbium (<0.0010) %/Thallium (<0.0010) %/Thorium (<0.0010) %/Thulium (<0.0010) %/Uranium (<0.0010) %/Ytterbium (<0.0010) %/Yttrium (<0.0010) %/Strontium (<0.0020) %/Bismuth (<0.0050) %/Cadmium (<0.0050) %/Hafnium (<0.0050) %/Rhenium (<0.0050) %/Lead (<0.0250) %/Calcium (0.0003) %/Germanium (0.001) %/Tantalum (0.001) %/Tungsten (0.002) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction