missing translation for 'onlineSavingsMsg'
Learn More

Nickel Ultrathin foil, 1.0 micron thick, 99.95% (metals basis excluding Co), Thermo Scientific Chemicals

Catalog Number 11358638
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
This item is not returnable. View return policy
7440-02-0
Ni
58.69
MFCD00011137 MFCD06798735
PXHVJJICTQNCMI-UHFFFAOYSA-N
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
935
CHEBI:28112
nickel
[Ni]
This item is not returnable. View return policy
7440-02-0
Ni
58.69
MFCD00011137 MFCD06798735
PXHVJJICTQNCMI-UHFFFAOYSA-N
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
935
CHEBI:28112
nickel
[Ni]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 25 x 25 mm
Percent Purity 99.95%
Odor Odorless
Particle Size 1.0 Micron thick
Assay (metals basis excluding Co)
Chemical Name or Material Nickel Ultrathin foil

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'