missing translation for 'onlineSavingsMsg'
Learn More

Nickel tungsten oxide, 99.9% (metals basis excluding Co), Co <100ppm

Product Code. 11394777
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394777 100 g 100g
11384777 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394777 Supplier Thermo Scientific Alfa Aesar Supplier No. 041826.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14177-51-6 | NiO4W | 306.529 g/mol

Nickel tungsten oxide is used as photo catalysts, sensors and humidity dielectric resonators. It is considered as a new cathode material for asymmetric super capacitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14177-51-6
Molecular Formula NiO4W
Molecular Weight (g/mol) 306.529
MDL Number MFCD00016268
InChI Key QLTKZXWDJGMCAR-UHFFFAOYSA-N
Synonym nickel tungstate, nickel tungsten tetraoxide, nickel tungsten oxide niwo4, nickel 2+ dioxido dioxo tungsten, nickel 2+ ion tungstate, dioxido dioxo tungsten; nickel 2+
PubChem CID 84240
IUPAC Name dioxido(dioxo)tungsten;nickel(2+)
SMILES [O-][W](=O)(=O)[O-].[Ni+2]

Specifications

Physical Form Powder
Quantity 100 g
Solubility Information Insoluble in water.
Formula Weight 306.53
Percent Purity 99.9%
Odor Odorless
Assay (metals basis excluding Co), Co <100ppm
Chemical Name or Material Nickel tungsten oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.