missing translation for 'onlineSavingsMsg'
Learn More

Nickel sulfide, 99.9% (metals basis)

Product Code. 11485040
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11485040 10 g 10g
11383807 50 g 50g
11393807 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11485040 Supplier Thermo Scientific Alfa Aesar Supplier No. 035661.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12035-72-2 | Ni3S2 | 240.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12035-72-2
Molecular Formula Ni3S2
Molecular Weight (g/mol) 240.20
MDL Number MFCD01711880
InChI Key YGHCWPXPAHSSNA-UHFFFAOYSA-N
Synonym nickel subsulfide, trinickel disulphide, nickel sulfide, trinickel disulfide, nickel subsulphide, nickel sesquisulfide, nickel tritadisulphide, nickel iii sulfide, alpha nickel subsulfide, ccris 432
PubChem CID 22833473
SMILES S=[Ni][Ni][Ni]=S

Specifications

Melting Point 797°C
Color Black
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.9% (metals basis)
Solubility Information Soluble in water(0,00735g/L at 20°C).
IUPAC Name disulfanylidenetrinickel
Formula Weight 240.19
Appearance Black powder
Chemical Name or Material Nickel sulfide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.