missing translation for 'onlineSavingsMsg'
Learn More

Nickel Sulfate Hexahydrate, TRC

Product Code. 30428571
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 30428571

Brand: Toronto Research Chemicals TRCN901063500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nickel(II) Sulfate Hexahydrate
CAS 10101-97-0
Molecular Formula 6 H2 O . Ni . O4 S
Quantity 500 mg
Synonym Nickel Sulfate Hexahydrate, Sulfuric acid','nickel(2+) salt','hydrate (1:1:6), Sulfuric acid','nickel(2+) salt (1:1)','hexahydrate (8CI,9CI), Nickel monosulfate hexahydrate, Nickel sulfate (NiSO4) hexahydrate, Nickel sulfate hexahydrate, Nickel sulfate hydrate (NiSO4.6H2O), Nickel(2+) monosulfate hexahydrate, Nickel(2+) sulfate hexahydrate, Nickel(II) sulfate hexahydrate
InChI Formula InChI=1S/Ni.H2O4S.6H2O/c;1-5(2,3)4;;;;;;/h;(H2,1,2,3,4);6*1H2/q+2;;;;;;;/p-2
SMILES O.O.O.O.O.O.[Ni+2].[O-]S(=O)(=O)[O-]
IUPAC Name nickel(2+);sulfate;hexahydrate
Molecular Weight (g/mol) 262.848
Formula Weight 261.95
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.