missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nickel sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.99% (metals basis)

Product Code. 15444815
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15444815 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15444815 Supplier Alfa Aesar™ Supplier No. 040877.KS
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 7440-02-0
Quantity 1 Ea.
Odor Odorless
Molecular Formula Ni
MDL Number MFCD00011137 MFCD06798735
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES [Ni]
IUPAC Name nickel
Molecular Weight (g/mol) 58.69
PubChem CID 935
ChEBI CHEBI:28112
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Nickel sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.