missing translation for 'onlineSavingsMsg'
Learn More

Nickel silicide, 99% (metals basis excluding Co), Co 0.1-1%, Thermo Scientific Chemicals

Product Code. 11383308
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383308 50 g 50g
11393308 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383308 Supplier Thermo Scientific Alfa Aesar Supplier No. 036251.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ni silicides have been widely investigated for contact materials in microelectronic devices. Electroplating is another major use of the metal. nickel silicide/silicon nanowire heterostructures have been used to produce field-effect transistors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12059-14-2
Molecular Formula Ni2Si
Molecular Weight (g/mol) 145.472
MDL Number MFCD00151367
InChI Key RUFLMLWJRZAWLJ-UHFFFAOYSA-N
Synonym nickel silicide, dinickelylidenesilane
PubChem CID 14767304
SMILES [Si](=[Ni])=[Ni]

Specifications

Physical Form Powder
Quantity 50 g
UN Number UN3077
Solubility Information Insoluble in water.
Formula Weight 145.47
Percent Purity 99%
Odor Odorless
Assay (metals basis excluding Co), Co 0.1 to 1%
Chemical Name or Material Nickel silicide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.