Learn More
Nickel silicide, 99% (metals basis excluding Co), Co 0.1-1%, Thermo Scientific Chemicals
Description
Ni silicides have been widely investigated for contact materials in microelectronic devices. Electroplating is another major use of the metal. nickel silicide/silicon nanowire heterostructures have been used to produce field-effect transistors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 250 g |
| UN Number | UN3077 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 145.47 |
| Percent Purity | 99% |
| Odor | Odorless |
| Assay | (metals basis excluding Co), Co 0.1 to 1% |
| Chemical Name or Material | Nickel silicide |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.