missing translation for 'onlineSavingsMsg'
Learn More

Nickel on silica-alumina, catalyst

Product Code. p-4408744
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11327448

Brand: Thermo Scientific Alfa Aesar 031276.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Formula: 66±5% Ni | CAS: 7440-02-0 | Ni | 58.69 g/mol

Nickel on Silica-Alumina Catalys products are often used as the most efficient way to catalyze organic reactions. It is also used in R&D, manufacture of chemicals, Pharmaceuticals and intermediates. It is an important raw material and intermediate used in Organic Synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-02-0
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

Color Black
Appearance Black catalyst powder
Quantity 5 g
Odor Odorless
Form Powder
Solubility Information It is insoluble in water.
Chemical Name or Material Nickel, on silica-alumina, catalyst
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Nickel on silica-alumina, catalyst
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Skin sensitiser Category 1
  • Carcinogenicity Category 1A
  • Specific target organ toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 4
Hazard Statement
  • H228-Flammable solid.
  • H317-May cause an allergic skin reaction.
  • H350i-May cause cancer by inhalation.
  • H372-Causes damage to organs.
  • H373-May cause damage to organs.
  • H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.