missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Reagent Set, Lovibond™
missing translation for 'orderingAttributeHoverText'
Quantity:
50 Tests
missing translation for 'unitSize'
Pack of 50
Specifications
Specifications
| Quantity | 50 Tests |
Safety and Handling
Product Identifier
- reagent test nickel
- Danger
- Skin corrosion/irritation Category 1 A
- Oxidising solid Category 3
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H314-Causes severe skin burns and eye damage.
- H272-May intensify fire; oxidiser.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H317-May cause an allergic skin reaction.
- H335-May cause respiratory irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P313-Get medical advice/attention.
- MIXTURE LIST-Contains : sodium hydroxide, dimethylglyoxime, ammonium persulphate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction