missing translation for 'onlineSavingsMsg'
Learn More

Nickel powder, spherical, -100+325 mesh, 99.8% (metals basis), Thermo Scientific™

Catalog Number 11375517
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
7440-02-0
Ni
58.69
MFCD00011137 MFCD06798735
PXHVJJICTQNCMI-UHFFFAOYSA-N
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
935
CHEBI:28112
nickel
[Ni]
This item is not returnable. View return policy
7440-02-0
Ni
58.69
MFCD00011137 MFCD06798735
PXHVJJICTQNCMI-UHFFFAOYSA-N
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
935
CHEBI:28112
nickel
[Ni]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Gray
UN Number UN3089
Quantity 50 g
Percent Purity 99.8%
Mesh Size −100+325 Mesh
Odor Odorless
Form Spherical Powder
Assay (metals basis)
Chemical Name or Material Nickel powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'