Learn More
Nickel powder, -325 mesh, typically 99.8% (metals basis), Thermo Scientific Chemicals
Form: Conductive dendritic powder
£17.70 - £400.00
Chemical Identifiers
CAS | 7440-02-0 |
---|---|
Molecular Formula | Ni |
Molecular Weight (g/mol) | 58.69 |
MDL Number | MFCD00011137 MFCD06798735 |
InChI Key | PXHVJJICTQNCMI-UHFFFAOYSA-N |
Synonym | raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum |
PubChem CID | 935 |
ChEBI | CHEBI:28112 |
IUPAC Name | nickel |
SMILES | [Ni] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11319288
|
Thermo Scientific Alfa Aesar
043214.14 |
25 g |
£17.70
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11399278
|
Thermo Scientific Alfa Aesar
043214.22 |
100 g |
£48.10
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11389278
|
Thermo Scientific Alfa Aesar
043214.36 |
500 g |
£131.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11309288
|
Thermo Scientific Alfa Aesar
043214.A3 |
2 kg |
£400.00
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Conductive surface coatings, inks, adhesives, and sealants
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-02-0 | |
58.69 | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
935 | |
nickel |
Ni | |
MFCD00011137 MFCD06798735 | |
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
CHEBI:28112 | |
[Ni] |
Specifications
Silver to Gray | |
Silver gray powder | |
Ni | |
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
[Ni] | |
58.69 | |
CHEBI:28112 | |
Odorless | |
Nickel |
7440-02-0 | |
99.8% (Metals basis) | |
MFCD00011137 MFCD06798735 | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
nickel | |
935 | |
-325 Mesh | |
Conductive Dendritic Powder |
Safety and Handling
Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). Avoid release to the environment. Refer to special instructions/Safety data sheets.
P201-P202-P210-P240-P241-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P370+P378q-P501c
H228-H317-H350-H372
missing translation for 'dotInformation' : DOT Class: 4.1, Packaging Group: II
missing translation for 'einecsNumber' : 231-111-4
missing translation for 'rtecsNumber' : QR5950000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only