Learn More
Bochem™ Nickel Plated Brass Pinch Cock
Used to control the flow of liquid through flexible tubing
Brand: Bochem™ 12534
Code : A0
Additional Details : Weight : 0.80000kg
Description
- With flap
- Manufactured from Nickel Plated Brass
Specifications
Hoffman Pinchcock | |
Nickel Plated Brass | |
30 mm |