Learn More
Cole-Parmer™ Nickel-plated Brass Luer Fittings
Product Code. 15392132
Click to view available options
For Use With:
Female luer lock
Male Luer Lock
Includes:
1/16 in. Hose Barb
10-32 UNF Thread
Unit Size:
Each
Product Code. 15392132
Brand: Cole-Parmer™ 4150762
Only null left
Add to basket
Add to basket
Specifications
Autoclavable Autoclavable
Material Nickle-plated Brass
For Use With Male Luer Lock
For Use With (Application) Harsh chemical environments
Packaging Quantity 1 Pack
Includes 1/16 in. Hose Barb
Product Type Luer Fitting