missing translation for 'onlineSavingsMsg'
Learn More

Nickel, plasma standard solution, Specpure™ Ni 1000μg/mL

Product Code. 11430350
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
100mL
500mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430350 500 mL 500mL
11331399 50 mL 50mL
11321399 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11430350 Supplier Thermo Scientific Alfa Aesar Supplier No. 013839.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-02-0 | Ni | 58.69 g/mol

Nickel, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-02-0 , 7697-37-2 , 7732-18-5
Molecular Weight (g/mol) 58.69
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Molecular Formula Ni
MDL Number MFCD00011137 MFCD06798735
Color Colorless
Solubility Miscible with water
Quantity 500 mL
Concentration or Composition (by Analyte or Components) Nitric acid: 5.0%; Nickel: 0.1%; Water: 94.9%
Chemical Name or Material Nickel

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.