missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel, Ni/Mo, Alloy B-2, N10665, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Nickel, Ni/Mo, Alloy B-2, N10665, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Magnesium/Boron/Zirconium/Copper/Phosphorus/Vanadium/Carbon/Cobalt/Tungsten/Silicon/Niobium/Aluminium/Titanium/Chromium/Manganese/Iron/Molybdenum/Nickel/Beryllium/Bismuth/Cerium/Germanium/Indium/Palladium/Ruthenium/Selenium/Tellurium/Zinc/Antimony/Sodium/Arsenic/Calcium/Lead/Silver/Yttrium/Oxygen/Hafnium/Lanthanum/Tantalum/Cadmium/Gallium/Rhenium/Nitrogen/Tin |
| CAS | 7440-02-0 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Magnesium 0.0013 %/Boron 0.0017 %/Zirconium 0.002 %/Copper 0.003 %/Phosphorus 0.004 %/Vanadium 0.007 %/Carbon 0.0079 %/Cobalt 0.013 %/Tungsten 0.02 %/Silicon 0.03 %/Niobium 0.04 %/Aluminium 0.13 %/Titanium 0.14 %/Chromium 0.153 %/Manganese 0.76 %/Iron 1.56 %/Molybdenum 26.6 %/Nickel 70.4 %/Beryllium (<0.0001) %/Bismuth (<0.0001) %/Cerium (<0.0001) %/Germanium (<0.0001) %/Indium (<0.0001) %/Palladium (<0.0001) %/Ruthenium (<0.0001) %/Selenium (<0.0002) %/Tellurium (<0.0003) %/Zinc (<0.0003) %/Antimony (<0.0005) %/Sodium (<0.0005) %/Arsenic (<0.001) %/Calcium (<0.001) %/Lead (<0.001) %/Silver (<0.001) %/Yttrium (<0.001) %/Oxygen (<0.006) %/Hafnium (<0.01) %/Lanthanum (<0.01) %/Tantalum (<0.011) %/Cadmium (<0.07) %/Gallium (0.0001) %/Rhenium (0.0001) %/Nitrogen (0.0007) %/Tin (0.002) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction