missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel, Ni/Cu, Alloy R405, N04405, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Nickel, Ni/Cu, Alloy R405, N04405, 100g
Specifications
Specifications
| Chemical Name or Material | Calcium/Vanadium/Boron/Tin/Silver/Tantalum/Antimony/Arsenic/Nitrogen/Oxygen/Lead/Tungsten/Lanthanum/Zirconium/Titanium/Phosphorus/Magnesium/Cobalt/Sulfur/Aluminium/Molybdenum/Niobium/Carbon/Silicon/Chromium/Manganese/Iron/Copper/Nickel/Hafnium/Cadmium/Bismuth |
| CAS | 7440-02-0 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Calcium -20 ppm/Vanadium -20 ppm/Boron -6 ppm/Tin -6 ppm/Silver -4 ppm/Tantalum -4 ppm/Antimony -2 ppm/Arsenic 0.0005 %/Nitrogen 0.0007 %/Oxygen 0.0008 %/Lead 0.001 %/Tungsten 0.005 %/Lanthanum 0.006 %/Zirconium 0.006 %/Titanium 0.0086 %/Phosphorus 0.011 %/Magnesium 0.018 %/Cobalt 0.04 %/Sulfur 0.04 %/Aluminium 0.043 %/Molybdenum 0.102 %/Niobium 0.103 %/Carbon 0.117 %/Silicon 0.161 %/Chromium 0.499 %/Manganese 0.96 %/Iron 1.71 %/Copper 31.2 %/Nickel 64.8 %/Hafnium (<100) ppm/Cadmium (<20) ppm/Bismuth (<50) ppm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction