missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel, Ni/Cu, Alloy 400, N04400, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Nickel, Ni/Cu, Alloy 400, N04400, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Niobium/Phosphorus/Cobalt/Magnesium/Molybdenum/Titanium/Aluminium/Chromium/Carbon/Silicon/Manganese/Iron/Copper/Nickel/Antimony/Beryllium/Cadmium/Cerium/Hafnium/Indium/Lanthanum/Palladium/Rhenium/Ruthenium/Tellurium/Boron/Selenium/Silver/Sodium/Yttrium/Zinc/Oxygen/Calcium/Lead/Tantalum/Germanium/Arsenic/Bismuth/Tin/Gallium/Nitrogen/Zirconium/Vanadium/Tungsten |
| CAS | 7440-02-0 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Niobium 0.006 %/Phosphorus 0.01 %/Cobalt 0.011 %/Magnesium 0.016 %/Molybdenum 0.019 %/Titanium 0.033 %/Aluminium 0.036 %/Chromium 0.064 %/Carbon 0.139 %/Silicon 0.16 %/Manganese 1.03 %/Iron 1.68 %/Copper 32.6 %/Nickel 64.1 %/Antimony (<0.0001) %/Beryllium (<0.0001) %/Cadmium (<0.0001) %/Cerium (<0.0001) %/Hafnium (<0.0001) %/Indium (<0.0001) %/Lanthanum (<0.0001) %/Palladium (<0.0001) %/Rhenium (<0.0001) %/Ruthenium (<0.0001) %/Tellurium (<0.0001) %/Boron (<0.0003) %/Selenium (<0.0004) %/Silver (<0.0004) %/Sodium (<0.0005) %/Yttrium (<0.0005) %/Zinc (<0.0005) %/Oxygen (<0.0007) %/Calcium (<0.001) %/Lead (<0.001) %/Tantalum (<0.001) %/Germanium (<0.002) %/Arsenic (<0.003) %/Bismuth (<0.003) %/Tin (0.00016) %/Gallium (0.00019) %/Nitrogen (0.00019) %/Zirconium (0.0008) %/Vanadium (0.001) %/Tungsten (0.0017) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction