missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel, Ni/Cr, Alloy 600, N06600, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Nickel, Ni/Cr, Alloy 600, N06600, 100g
Specifications
Specifications
| Chemical Name or Material | Tin/Boron/Tantalum/Phosphorus/Tungsten/Magnesium/Vanadium/Cobalt/Carbon/Copper/Molybdenum/Niobium/Aluminium/Silicon/Titanium/Manganese/Iron/Chromium/Nickel/Beryllium/Bismuth/Cerium/Hafnium/Indium/Lanthanum/Palladium/Rhenium/Ruthenium/Silver/Tellurium/Yttrium/Cadmium/Sodium/Germanium/Arsenic/Calcium/Selenium/Zinc/Antimony/Lead/Oxygen/Gallium/Zirconium/Nitrogen |
| CAS | 7440-02-0 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Tin 0.002 %/Boron 0.0026 %/Tantalum 0.004 %/Phosphorus 0.007 %/Tungsten 0.014 %/Magnesium 0.016 %/Vanadium 0.024 %/Cobalt 0.056 %/Carbon 0.077 %/Copper 0.077 %/Molybdenum 0.084 %/Niobium 0.088 %/Aluminium 0.17 %/Silicon 0.17 %/Titanium 0.255 %/Manganese 0.26 %/Iron 9.5 %/Chromium 16 %/Nickel 72.7 %/Beryllium (<0.0001) %/Bismuth (<0.0001) %/Cerium (<0.0001) %/Hafnium (<0.0001) %/Indium (<0.0001) %/Lanthanum (<0.0001) %/Palladium (<0.0001) %/Rhenium (<0.0001) %/Ruthenium (<0.0001) %/Silver (<0.0001) %/Tellurium (<0.0001) %/Yttrium (<0.0001) %/Cadmium (<0.0003) %/Sodium (<0.0005) %/Germanium (<0.0008) %/Arsenic (<0.001) %/Calcium (<0.001) %/Selenium (<0.001) %/Zinc (<0.002) %/Antimony (0.00011) %/Lead (0.0006) %/Oxygen (0.001) %/Gallium (0.0016) %/Zirconium (0.002) %/Nitrogen (0.007) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction