missing translation for 'onlineSavingsMsg'
Learn More

Nickel Metal Powder, SLR, Extra Pure, Fisher Chemical

Product Code. 10090290 Shop All Fisher Chemical Products
Click to view available options
Quantity:
250 g
Packaging:
Plastic powder jar
Unit Size:
250g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10090290

Brand: Fisher Chemical N/1300/50

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Specifications

Color Brown
CAS 7440-02-0
Melting Point 1455°C
Boiling Point 2730°C
Quantity 250 g
Packaging Plastic powder jar
Vapor Pressure 1mmHg 1810
Form Solid
Molecular Formula Ni
MDL Number MFCD00011137 MFCD06798735
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES [Ni]
IUPAC Name nickel
Molecular Weight (g/mol) 58.69
PubChem CID 935
ChEBI CHEBI:28112
Formula Weight 58.7g/mol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Nickel, powder
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
  • H351-Suspected of causing cancer.
  • H412-Harmful to aquatic life with long lasting effects.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.