missing translation for 'onlineSavingsMsg'
Learn More

Nickel Isotopic Standard: Ni @ 10 μg/mL in 2% HNO3 (Enriched in Ni-61), VHG™

Product Code. 30003122
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003122 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30003122 Supplier VHG Labs Supplier No. VHGLIS61NI50

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Nickel Isotopic Standard: Ni @ 10 μg/mL in 2% HNO3 (Enriched in Ni-61)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nickel 61
CAS 14928-10-0
Methods ICP-MS
Analyte or Component Names Nickel 61
Shipping Condition Room Temperature
Molecular Formula 61Ni
Synonym 61Ni
SMILES [61Ni]
IUPAC Name Nickel 61
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 60.93
Quantity 50 mL
Formula Weight 60.9311 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.