Learn More
Nickel Iron, sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis), Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 040888.KS
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nickel Iron, sputtering target | |
1 Ea. | |
MFCD01741928 |
76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick | |
(metals basis) | |
99.95% |
Safety and Handling
- Nickel Iron, sputtering target
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
- Carcinogenicity Category 2
Hazard Statement
- H317-May cause an allergic skin reaction.
- H351-Suspected of causing cancer.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P363-Wash contaminated clothing before reuse.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only